Skip to Content
Merck
All Photos(1)

Key Documents

SCP0229

Sigma-Aldrich

Protein Tyrosine Phosphatase Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€262.00

€262.00


Usually ships in 2 weeks.


Select a Size

Change View
1 MG
€262.00

About This Item

Empirical Formula (Hill Notation):
C59H77N16O25
Molecular Weight:
1410.33
UNSPSC Code:
12352202
NACRES:
NA.32

€262.00


Usually ships in 2 weeks.

Product Name

Protein Tyrosine Phosphatase Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

MCA-Gly-Asp-Ala-Glu-pTyr-Ala-Ala-DNP-Lys-Arg-NH2

Application

Protein Tyrosine Phosphatase Substrate (MCA-Gly-Asp-Ala-Glu-pTyr-Ala-Ala-DNP-Lys-Arg-NH2) may be used to identify and differentiate protein tyrosine phosphatases.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Klein et al.
Scientific reports, 10(1), 15522-15522 (2020-09-25)
Enzymes are fundamental to biological processes and involved in most pathologies. Here we demonstrate the concept of simultaneously mapping multiple enzyme activities (EA) by applying enzyme substrate libraries to tissue sections and analyzing their conversion by matrix-assisted laser desorption/ionization (MALDI)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service