Skip to Content
Merck
All Photos(1)

Key Documents

SCP0221

Sigma-Aldrich

Procaspase 3 Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€186.00

€186.00


Usually ships in 2 weeks.


Select a Size

Change View
5 MG
€186.00

About This Item

Empirical Formula (Hill Notation):
C29H34N5O12S1
Molecular Weight:
676.67
UNSPSC Code:
12352200
NACRES:
NA.32

€186.00


Usually ships in 2 weeks.

Product Name

Procaspase 3 Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥77%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Glu-Ser-Met-Asp-AFC

Application

Procaspase 3 Substrate (Ac-ESMD-AFC) contains the target sequence for the proteolytic production of the p17 subunit of caspase 3 from its precursor (procaspase). Ac-ESMD-CHO inhibits this activity. Ac-ESMD-AFC may be used in comparison with other procaspase substrate and inhibitor peptides to differentiate proteolytic activity used for procaspase processing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominique Le Goff et al.
Reproductive toxicology (Elmsford, N.Y.), 21(3), 329-334 (2005-11-02)
The aim of the present study was to determine whether or not apoptosis occurs in Sertoli cells in presence of 25-hydroxycholesterol, an oxysterol derived from cholesterol-containing foods or endogenous oxidation. Here, we provide evidence that 25-hydroxycholesterol can induce cultured Sertoli
Johnson Agniswamy et al.
The FEBS journal, 274(18), 4752-4765 (2007-08-19)
Many protein substrates of caspases are cleaved at noncanonical sites in comparison to the recognition motifs reported for the three caspase subgroups. To provide insight into the specificity and aid in the design of drugs to control cell death, crystal
Z Han et al.
The Journal of biological chemistry, 272(20), 13432-13436 (1997-05-16)
The apoptotic cysteine protease, caspase-3, is expressed in cells as an inactive 32-kDa precursor from which 17 kDa (p17) and 12 kDa (p12) subunits of the mature caspase-3 are proteolytically generated during apoptosis. Two amino acid sequences, ESMD downward arrowS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service