Skip to Content
Merck
All Photos(1)

Key Documents

SCP0119

Sigma-Aldrich

Vasopressin Peptide

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€255.00

€255.00


Usually ships in 2 weeks.


Select a Size

Change View
5 MG
€255.00

About This Item

Empirical Formula (Hill Notation):
C48H69N13O11S2
Molecular Weight:
1068.27
UNSPSC Code:
12352209
NACRES:
NA.32

€255.00


Usually ships in 2 weeks.

Product Name

[Deamino-Pen1,Val4,D-Arg8]-Vasopressin,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥73%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Deamino-Pen-Tyr-Phe-Val-Asn-Cys-Pro-Arg-Gly-NH2

Application

Arginine vasopressin (AVP) is a neurohypophysial hormone involved in the regulation of water retension and blood pressure. [Deamino-Pen1,Val4,D-Arg8]-Vasopressin is used as a peptide AVP antagonist versus non-peptide AVP antagonists such as tolvaptan.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service