Skip to Content
Merck
All Photos(1)

Key Documents

SCP0116

Sigma-Aldrich

Chromogranin A (324-337) WE 14 human

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€186.00

€186.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
€186.00

About This Item

Empirical Formula (Hill Notation):
C72H116N18O24S1
Molecular Weight:
1649.86
UNSPSC Code:
12352200
NACRES:
NA.32

€186.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥68%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Trp-Ser-Lys-Met-Asp-Gln-Leu-Ala-Lys-Glu-Leu-Thr-Ala-Glu

Application

Chromogranin A (parathyroid secretory protein 1) is a precursor of the vasostatin, pancreastatin, catestatin, parastatin, chromostatin, GE-25 and WE-14. WE 14 is used to search for its as yet undefined physiological roles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Per-Gøran Krüger et al.
Regulatory peptides, 114(1), 29-35 (2003-05-24)
Catestatin (bovine CgA(344-364)) is a cationic peptide, which besides reducing catecholamine secretion from chromaffin cells in vitro also acts a potent vasodilator in the rat in vivo. The alleged histamine releasing effect of catestatin was tested in vitro in rat
W J Curry et al.
Annals of the New York Academy of Sciences, 971, 311-316 (2002-11-20)
The neuropeptide WE-14 is derived from the posttranslational processing of chromogranin A (CgA). While CgA is expressed in a preponderance of neuroendocrine cells, WE-14 is generated in a distinct subpopulation of CgA-immunopositive cells, most notably in the adrenal, pituitary, and
The modulatory effects of WE-14 on histamine release from rat peritoneal mast cells.
P Forsythe et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 46 Suppl 1, S13-S14 (1997-03-01)
Carmel Mary McVicar et al.
Regulatory peptides, 115(1), 1-10 (2003-07-23)
Immunohistochemical analysis of the male and female rat adenohypophysis revealed that chromogranin A (CgA), beta-granin and WE-14 immunostaining was localised to follicle stimulating hormone (FSH) producing cells, while luteinizing hormone (LH) producing cells exhibited chromogranin A and beta-granin immunostaining. The
William James Curry et al.
Microscopy research and technique, 62(5), 408-414 (2003-11-06)
WE-14 is generated in subpopulations of chromogranin A immunopositive endocrine cells and neurons including those innervating the anterior uvea. This study investigated WE-14 in intact sclero-limbo-corneal tissue from embryonic (E17), neonatal (N0-N16), and adult mice using immunocytochemistry and confocal scanning

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service