Skip to Content
Merck
All Photos(1)

Key Documents

SCP0078

Sigma-Aldrich

Caspase 2 Substrate (ICH-1), fluorogenic

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€186.00

€186.00


Usually ships in 2 weeks.


Select a Size

Change View
5 MG
€186.00

About This Item

Empirical Formula (Hill Notation):
C58H71N13O20
Molecular Weight:
1270.26
UNSPSC Code:
12352202
NACRES:
NA.32

€186.00


Usually ships in 2 weeks.

Product Name

Caspase 2 Substrate (ICH-1), fluorogenic,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Related Categories

Amino Acid Sequence

MCA-Val-Asp-Val-Ala-Asp-Gly-Trp-DNP-Lys-NH2

Application

Caspase 2 is a cysteine protease that is potentially involved in cell cycle regulation and tumour suppression may be detected using peptides that contain the pentapeptide VDVAD such as the chromogenic and fluorogenic substrates: Ac-VDVAD-pNa (Acetyl-Val-Asp-Val-Ala-Asp-p- nitroanalide), chromogenic and MCA-VDVADGW-DNP-K-NH2, fluorogenic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanja Kitevska et al.
Apoptosis : an international journal on programmed cell death, 14(7), 829-848 (2009-05-30)
The caspases are an evolutionarily conserved family of cysteine proteases, with essential roles in apoptosis or inflammation. Caspase-2 was the second caspase to be cloned and it resembles the prototypical nematode caspase CED-3 more closely than any other mammalian protein.
Yinyan Tang et al.
The Journal of biological chemistry, 286(39), 34147-34154 (2011-08-11)
Caspase-2, the most evolutionarily conserved member in the human caspase family, may play important roles in stress-induced apoptosis, cell cycle regulation, and tumor suppression. In biochemical assays, caspase-2 uniquely prefers a pentapeptide (such as VDVAD) rather than a tetrapeptide, as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service