Skip to Content
Merck
All Photos(1)

Key Documents

SCP0022

Sigma-Aldrich

ANP [Des18-22] 4-23 Amide rat

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€246.00

€246.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
€246.00

About This Item

Empirical Formula (Hill Notation):
C64H107N25O19S2
Molecular Weight:
1594.82
UNSPSC Code:
51111800
NACRES:
NA.32

€246.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Related Categories

Amino Acid Sequence

Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Cys-NH2

Application

Atrial natriuretic peptide (ANP) is a 28 amino acid polypeptide vasodilator involved in water, sodium, potassium homeostasis. ANP mediates its actions via three receptor types: two guanylyl cyclase receptors know as NPR1 and NPR2 and a natriuretic peptide clearance receptor (NPR3/NPRC). ANP (4-23) is a selective agonist of natriuretic peptide receptor subtype NPR-C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service