Skip to Content
Merck
All Photos(1)

Key Documents

SCP0005

Sigma-Aldrich

MG-115 Peptide

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€193.00

€193.00


Usually ships in 2 weeks.


Select a Size

Change View
5 MG
€193.00

About This Item

Empirical Formula (Hill Notation):
C25H39N3O5
Molecular Weight:
461.59
UNSPSC Code:
12352202
NACRES:
NA.32

€193.00


Usually ships in 2 weeks.

Product Name

MG-115 / ASD 4955,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Z-Leu-Leu-Nva-al

Application

MG-115 (Z-Leu-Leu-Nva-al) is used as a selective proteosome inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yolanda Pérez-Pertejo et al.
Parasitology, 138(9), 1082-1092 (2011-08-05)
Methionine adenosyltransferase (MAT) is an important enzyme for metabolic processes, to the extent that its product, S-adenosylmethionine (AdoMet), plays a key role in trans-methylation, trans-sulphuration and polyamine synthesis. Previous studies have shown that a MAT-overexpressing strain of Leishmania donovani controls
Estabelle Ang et al.
Journal of cellular physiology, 220(2), 450-459 (2009-04-15)
Proteasome inhibitors represent a promising therapy for the treatment of relapsed and/or refractory multiple myeloma, a disease that is concomitant with osteolysis and enhanced osteoclast formation. While blockade of the proteosome pathway has been recently shown to influence osteoclast formation
Karim Harhouri et al.
EMBO molecular medicine, 9(9), 1294-1313 (2017-07-05)
Hutchinson-Gilford progeria syndrome (HGPS) is a lethal premature and accelerated aging disease caused by a de novo point mutation in LMNA encoding A-type lamins. Progerin, a truncated and toxic prelamin A issued from aberrant splicing, accumulates in HGPS cells' nuclei

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service