Skip to Content
Merck
All Photos(1)

Key Documents

SBR00121

Amphotericin B water soluble

new

from semisynthetic, powder

Synonym(s):

AmB, Amphotericin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H82N2O19
Molecular Weight:
1027.20
UNSPSC Code:
51302401
Pricing and availability is not currently available.

biological source

fermentation/recombinant
semisynthetic

Quality Level

Assay

≥90% (HPLC)

form

powder

storage condition

protect from light

color

light yellow to very dark yellow, to very dark brown

Mode of action

cell membrane | interferes

shipped in

dry ice

storage temp.

−20°C

suitability

fungi
yeasts

SMILES string

C[C@@H](O1)[C@@H](O)[C@H]([NH3+])[C@H](O)[C@]1([H])O[C@H](C[C@@]2([H])[C@@H]([C@@H](O)C[C@@](O2)(O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)C3)C(NCCO)=O)/C=C/C=C/C=C/C=C/C=C/C=C/C=C/[C@H](C)[C@@H](O)[C@@H](C)[C@H](C)OC3=O.CC([O-])=O

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5+,9-7+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-INPOYWNPSA-N

General description

Amphotericin B is a polyene antifungal antibiotic which has a high affinity for sterols, especially ergosterols, of both fungal and bacterial cell membranes.[1] Amphotericin B water soluble is a synthetic derivative of AmB that preserves the antifungal activity but is highly miscible in water up to 100 mg/mL.

Biochem/physiol Actions

Mode of Action: Amphotericin B binds to sterols, forming pores in the membrane, and causing small molecules to leak out. Antimicrobial spectrum: Includes fungi and yeast It has a high affinity for sterols, especially ergosterols, of both fungal and bacterial cell membranes. It forms transmembrane channels by binding to sterols associated with membranes, thereby leading to the leakage of small molecules[2] Amphotericin B elicits toxicity despite being an effective antibiotic for treating mycoses[3] Normal usage in maintenance of antifungal cell cultures is 2.5 mg/L with penicillin and streptomycin used in the medium. Amphotericin B is most effective against fungi and yeast.[2]

Features and Benefits

Easily soluble in water up to 100mg/mL
Animal component free
Tested for cell toxicity
Can be 0.2µm filtered

Preparation Note

Amphotericin B water soluble can be directly dissolved in the media to a working concentration of 2.5 mg/L.

Other Notes

Product is sensitive to light.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service