Skip to Content
Merck
All Photos(1)

Key Documents

SBR00031

Sigma-Aldrich

MIT HCl Ready Made Solution

(10 mg MIT/mL)

Synonym(s):

2-Methyl-1,2-thiazol-3-one, Methylisothiazolinone HCl, Methylisothiazolinone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51102829
NACRES:
NA.76

form

liquid

Quality Level

concentration

(10 mg MIT/mL)
1%

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

InChI

1S/C4H5NOS.ClH/c1-5-4(6)2-3-7-5;/h2-3H,1H3;1H

InChI key

SJXPQSRCFCPWQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methylisothiazolinone, also known as 2-Methyl-4-isothiazolin-3-one, belongs to the group of isothiazolinones. 2-Methyl-4-isothiazolin-3-one is utilized in cell biology and biochemical applications to effectively prevent the growth of bacterial contaminants, ensuring the purity of cell lines.

Application

Methylisothiazolinone (MIT) has been used:
  • in research on the effects of the N-methyl D-aspartate (NMDA) receptor antagonist kynurenic acid on human cortical development [1]
  • as a cytotoxic substance to investigate its effect on bronchial epithelial cells (BEAS-2B cells) and its role in apoptotic cell death
  • to research the effects of tyrosine phosphorylation on focal adhesion kinase (FAK) activity in the development of neural axons and dendrites

Biochem/physiol Actions

Mode of Action: Inhibits the growth of microorganisms by disrupting the normal function of microbial cells and interfering with essential cellular processes

Features and Benefits

  • Ready available solution reduce the need for preparation time
  • Commonly used in Cell Biology and Biochemical applications
  • High quality antibiotic suitable for mulitple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengxi Wei et al.
Toxicology letters, 338, 67-77 (2020-12-09)
Chemical-peptide conjugation is the molecular initiating event in skin sensitization. The OECD test guideline uses a high-performance liquid chromatography/ultraviolet (HPLC/UV) detection method to quantify chemical-peptide conjugation in a direct peptide reactivity assay (DPRA), which measures the depletion of two synthetic
Shen Du et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(17), 7408-7416 (2002-08-28)
Neurodegenerative disorders in humans may be triggered or exacerbated by exposure to occupational or environmental agents. Here, we show that a brief exposure to methylisothiazolinone, a widely used industrial and household biocide, is highly toxic to cultured neurons but not
Eun-Jung Park et al.
Toxicology in vitro : an international journal published in association with BIBRA, 62, 104661-104661 (2019-10-21)
Methylisothiazolinone (MIT) has been used in wide spectrum of fields due to its ability to inhibit microbial proliferation with low toxicity. Meanwhile, in Korea, the concern about the hazardous effects of MIT was amplified by the occurrence of patients that
Inseyah Bagasrawala et al.
Frontiers in neuroscience, 10, 435-435 (2016-10-18)
Kynurenic acid (KYNA), a neuroactive metabolite of tryptophan degradation, acts as an endogenous N-methyl-D-aspartate receptor (NMDAR) antagonist. Elevated levels of KYNA have been observed in pregnant women after viral infections and are considered to play a role in neurodevelopmental disorders.
M D Lundov et al.
The British journal of dermatology, 165(6), 1178-1182 (2011-07-23)
In the early 2000s the preservative methylisothiazolinone (MI) was released as an individual preservative for industrial products and, in 2005, it was permitted for use in cosmetic products. Up until then MI had been used only in combination with methylchloroisothiazolinone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service