Skip to Content
Merck
All Photos(1)

Key Documents

SBR00022

Sigma-Aldrich

Blasticidin S Ready Made Solution

10 mg/mL (20 mM HEPES), 0.22 μm filtered

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€79.60
10 ML
€733.00

€79.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€79.60
10 ML
€733.00

About This Item

Empirical Formula (Hill Notation):
C17H27N8O5Cl
Molecular Weight:
458.90
UNSPSC Code:
12352106
NACRES:
NA.76

€79.60


Check Cart for Availability

Request a Bulk Order

Quality Level

sterility

0.22 μm filtered

Assay

≥98% (HPLC)

form

liquid

concentration

10 mg/mL (20 mM HEPES)

color

colorless to faint yellow

antibiotic activity spectrum

fungi

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl

InChI

1S/C17H26N8O5.ClH/c1-24(16(20)21)6-4-9(18)8-12(26)22-10-2-3-13(30-14(10)15(27)28)25-7-5-11(19)23-17(25)29;/h2-3,5,7,9-10,13-14H,4,6,8,18H2,1H3,(H3,20,21)(H,22,26)(H,27,28)(H2,19,23,29);1H

InChI key

YQXYQOXRCNEATG-UHFFFAOYSA-N

General description

Blasticidin S, originally isolated from S. griseochromogenes, is a bacterial metabolite renowned for its antibacterial and fungicidal activities. This compound serves as a potent inhibitor of protein synthesis, exhibiting activity against various microorganisms, including bacteria (B. subtilis, S. lutea, E. coli, P. fluorescens, and M. tuberculosis), tumor cell lines, and nematodes.

In research, Blasticidin S has become a valuable tool, notably as a marker for strain manipulations. Recent applications involve the use of Blasticidin S as a selection agent for cells carrying plasmids conferring blasticidin resistance. The resistance is mediated by the blasticidin S deaminase genes (bsr from Bacillus cereus or BSD from Aspergillus terreus). These genes produce enzymes that catalyze the hydrolytic deamination of the cytosine moiety in blasticidin S, leading to the formation of a non-toxic deaminohydroxy derivative. This resistance mechanism is crucial in various studies involving genetic manipulations and selections in cell biology and biochemical research.

Application

Blasticidin S has been:
  • studies to have fungicidal properties and prevents rice blast disease.[1]
  • used as a selection agent for transformed cells that contain the resistance genes bls, bsr, or bsd. Blasticidin S has been used to select HEK293-T cells with TLR-2 constructs and HEK-D5 cells.[2][3]
  • used to study protein synthesis at the level of peptide bond formation.

Biochem/physiol Actions

Mode of Action: It inhibits protein synthesis in bacteria and eukaryotes.[4] Blasticidin S inhibits hydrolysis of peptidyl-tRNA induced by release factors. It enhances the binding of tRNA to the large subunit of ribosomes and to an extent inhibits peptide bond formation.[4][5]

Antimicrobial Spectrum: Active against mycobacteria, several Gram-positive and Gram-negative bacteria

Features and Benefits

  • High-quality antibiotic suitable for multiple research applications
  • Ideal for Cell Biology and Biochemical research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suneesh Kumar Pachathundikandi et al.
PloS one, 6(5), e19614-e19614 (2011-05-17)
Helicobacter pylori is the causative agent for developing gastritis, gastric ulcer, and even gastric cancer. Virulent strains carry the cag pathogenicity island (cagPAI) encoding a type-IV secretion system (T4SS) for injecting the CagA protein. However, mechanisms of sensing this pathogen
Egor Svidritskiy et al.
Journal of molecular biology, 430(5), 591-593 (2018-01-26)
Understanding the mechanisms of inhibitors of translation termination may inform development of new antibacterials and therapeutics for premature termination diseases. We report the crystal structure of the potent termination inhibitor blasticidin S bound to the ribosomal 70S•release factor 1 (RF1)
Martha Triantafilou et al.
PloS one, 8(4), e61199-e61199 (2013-04-18)
Ureaplasma species are the most frequently isolated microorganisms inside the amniotic cavity and have been associated with spontaneous abortion, chorioamnionitis, premature rupture of the membranes (PROM), preterm labour (PL) pneumonia in neonates and bronchopulmonary dysplasia in neonates. The mechanisms by
Egor Svidritskiy et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(30), 12283-12288 (2013-07-05)
The antibiotic blasticidin S (BlaS) is a potent inhibitor of protein synthesis in bacteria and eukaryotes. We have determined a 3.4-Å crystal structure of BlaS bound to a 70S⋅tRNA ribosome complex and performed biochemical and single-molecule FRET experiments to determine
Rohini Garg et al.
Journal of immunology (Baltimore, Md. : 1950), 184(11), 5980-5987 (2010-05-05)
Vi capsular polysaccharide is a major virulence determinant of the human typhoid- causing pathogen Salmonella typhi; it is absent in nontyphoidal Salmonella serovars. We show in this study that through its specific interaction with the membrane recognition complex containing the

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Can you please advise the freeze/thaw cycle of this product?

    1 answer
    1. This product is shipped at ambient temperature and may be stored at 4 °C for up to two weeks. For long term storage, -20 °C is recommended. The solution should be aliquoted prior to freezer storage. Avoid repeated freeze/thaw cycles.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service