Skip to Content
Merck
All Photos(1)

Key Documents

SBR00014

Sigma-Aldrich

Daptomycin Ready Made Solution

1 mg/mL in DMSO

Synonym(s):

N-decanoyl-L-tryptophyl-L-asparaginyl-L-aspartyl-L-threonylglycyl-L-ornithyl-L-aspartyl-D-alanyl-L-aspartylglycyl-D-seryl-threo-3-methyl-L-glutamyl-3-anthraniloyl-L-alanine[egr]1-lactone

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
€256.00

€256.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.5 ML
€256.00

About This Item

Empirical Formula (Hill Notation):
C72H101N17O26
CAS Number:
Molecular Weight:
1620.67
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.76

€256.00


Check Cart for Availability

Request a Bulk Order

biological source

Streptomyces roseosporus

Quality Level

description

Ready Made Solution

form

liquid

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

concentration

1 mg/mL in DMSO

color

colorless to faint yellow

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

DNA synthesis | interferes
protein synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(NCC(N[C@H](CCCN)C(N[C@@H](CC(O)=O)C(N[C@H](C)C(N[C@@H](CC(O)=O)C(NC1)=O)=O)=O)=O)=O)[C@@H](NC([C@H](CC(O)=O)NC([C@@H](CC(N)=O)NC([C@H](CC2=CNC3=C2C=CC=C3)NC(CCCCCCCCC)=O)=O)=O)=O)[C@@H](C)OC([C@H](CC(C4=C(N)C=CC=C4)=O)NC([C@@]([C@H](C)CC(O)=O)([H])NC(

InChI

1S/C72H101N17O26/c1-5-6-7-8-9-10-11-22-53(93)81-44(25-38-31-76-42-20-15-13-17-39(38)42)66(108)84-45(27-52(75)92)67(109)86-48(30-59(102)103)68(110)89-61-37(4)115-72(114)49(26-51(91)40-18-12-14-19-41(40)74)87-71(113)60(35(2)24-56(96)97)88-69(111)50(34-90)82-55(95)32-77-63(105)46(28-57(98)99)83-62(104)36(3)79-65(107)47(29-58(100)101)85-64(106)43(21-16-23-73)80-54(94)33-78-70(61)112/h12-15,17-20,31,35-37,43-50,60-61,76,90H,5-11,16,21-30,32-34,73-74H2,1-4H3,(H2,75,92)(H,77,105)(H,78,112)(H,79,107)(H,80,94)(H,81,93)(H,82,95)(H,83,104)(H,84,108)(H,85,106)(H,86,109)(H,87,113)(H,88,111)(H,89,110)(H,96,97)(H,98,99)(H,100,101)(H,102,103)/t35?,36-,37?,43+,44+,45+,46+,47+,48+,49+,50-,60+,61?/m1/s1

InChI key

DOAKLVKFURWEDJ-OFNKPWESSA-N

Biochem/physiol Actions

Daptomycin is a natural cyclic lipopeptide antibiotic found in Streptomyces roseosporus. It is has been found to be effective in treating infections caused by aerobic Gram-positive bacteria. Daptomycin inhibits many antibiotic resistant strains such as methicillin-resistant S. aureus (MRSA), vancomycin-intermediate S. aureus (VISA) and vancomycin-resistant S. aureus (VRSA).[1][2]

The mode of action of daptomycin is quite different from that of any other antibiotic. Daptomycin binds to bacterial membranes and causes a rapid depolarization of membrane potential. This loss of membrane potential causes inhibition of protein, DNA, and RNA synthesis, which results in bacterial cell death.[3][4]

Packaging

0.5ML

Preparation Note

The 1 mg/mL solution may be further diluted in an appropriate buffer prior to use. It is recommended that a fresh dilution be made on the day of use.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced pulmonary surfactant interaction of daptomycin analogs via tryptophan replacement with alternative amino acids.
He Y, et al.
Bioorganic & Medicinal Chemistry Letters, 22, 6248-6251 (2012)
[Daptomycin: revitalizing a former drug due to the need of new active agents against grampositive multiresistant bacterias].
Hernandez Marti V, et al.
Revista Espanola de Quimioterapia : Publicacion Oficial de la Sociedad Espanola de Quimioterapia, 20, 261-276 (2007)
Feng Wang et al.
Gene, 544(2), 208-215 (2014-04-29)
Daptomycin, a novel cyclic lipopeptide antibiotic against Gram-positive bacteria, is produced by Streptomyces roseosporus. Though its biosynthetic mechanism, structural shuffling and fermentation optimization have been extensively studied, little is understood about its production regulation at the transcriptional levels. Here we
Diixa Patel et al.
International journal of antimicrobial agents, 38(5), 442-446 (2011-08-16)
Daptomycin is highly active against Staphylococcus aureus, including multidrug-resistant strains and those with reduced susceptibility to vancomycin. However, daptomycin-non-susceptible (Dap(NS)) strains [minimum inhibitory concentration (MIC) >1mg/L] have been derived clinically and in vitro. The mechanism(s) by which this occurs is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service