Skip to Content
Merck
All Photos(1)

Key Documents

SAE0111

Sigma-Aldrich

Phosphatase, Alkaline from human placenta

≥10 units/mg solid (in glycine buffer)

Synonym(s):

Phosphatase, Alkaline from human placenta, Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing

Select a Size

100 UNITS
€124.00
1000 UNITS
€521.00

€124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 UNITS
€124.00
1000 UNITS
€521.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€124.00


Check Cart for Availability

Request a Bulk Order

biological source

human placenta

specific activity

≥10 units/mg solid (in glycine buffer)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ALPP(250)

Looking for similar products? Visit Product Comparison Guide

Application

Alkaline phosphatase is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection. It is routinely used to dephosphorylate proteins and nucleic acids. It may be used for protein labeling when high sensitivity is required. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase.

Biochem/physiol Actions

Alkaline phosphatase dephosphorylates proteins and nucleic acids. Human placental alkaline phosphatase (PLAP) has an unique residue, Glu-429. It is an allosteric enzyme that is uncompetitively inhibited by L-Phe, L-Trp, and L-Leu. PLAP is post-translationally modified. Phosphatidylinositol-specific phospholipase C may be involved in the release of PLAP. PLAP may have a role in cell division and may be involved in the transfer of maternal IgG to the fetus.

Unit Definition

One unit will hydrolyze 1.0 umole of P-Nitrophenyl Phosphate per minute at pH 10.4 at 37 Deg C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service