Skip to Content
Merck
All Photos(1)

Key Documents

SAE0099

Sigma-Aldrich

S-(5′-Adenosyl)-L-homocysteine ready made solution

≥98% (HPLC), 1 mg/ml in 0.02 M HCl

Synonym(s):

5′-Deoxy-S-adenosyl-L-homocysteine, AdoHcy, S-(5′-Deoxyadenosine-5′)-L-homocysteine, SAH

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€233.00

€233.00


Check Cart for Availability


Select a Size

Change View
1 ML
€233.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.51

€233.00


Check Cart for Availability

Assay

≥98% (HPLC)

concentration

1 mg/mL

shipped in

dry ice

storage temp.

−20°C

General description

S-(5′-Adenosyl)-L-homocysteine (SAH) is a compound that occurs biologically as a by-product of reactions which S-adenosyl-L-methionine-dependent methyltransferases catalyze. These enzymes use S-adenosyl-L-methionine (SAM) as the parent material for cellular methyl transfer processes, such as DNA methylation. In its turn, SAH is cleaved by SAH hydrolase to produce homocysteine and adenosine. The homocysteine pathway and the plasma measurement of SAM and SAH3 have been studied in relation to topics such as cancer, nutritional deficiencies, and Alzheimer′s disease. SAH has been used to develop various methyltransferase activity assays, such as the following:
  • Multiwell plate assay for protein methyltransferase SET7/9 activity
  • Fluorescence polarization (FP) assay
  • HPLC assay coupled to fluorometric detection

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service