Skip to Content
Merck
All Photos(1)

Key Documents

SAE0009

Sigma-Aldrich

Proteinase K from Tritirachium album

≥30 units/mg protein

Synonym(s):

Endopeptidase K

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€70.50
100 MG
€165.00
1 G
€952.00

€70.50


Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0005


Select a Size

Change View
25 MG
€70.50
100 MG
€165.00
1 G
€952.00

About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

€70.50


Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0005

biological source

fungus (Tritirachium album)

Quality Level

form

lyophilized powder

specific activity

≥30 units/mg protein

mol wt

28.93 kDa

technique(s)

DNA extraction: suitable

foreign activity

DNAse, RNAse, none detected.

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Proteinase K (PK) from fungi, Tritirachium album encodes a 40 kDa protein. The N-terminal propeptide region shows homology with bacterial subtilisin.[1] PK crystallizes even under microgravity conditions on the space shuttle mission.[2] PK is an effective protein system for studying protein engineering process.[3]

Application

Protease footprinting by Proteinase K digestion can reveal protein-protein surface interactions. The enzyme from Sigma has been used in the pre-hybridization step of chicken embryos. It has also been used for the enrichment of PrPSc, a prion protein that is present in sheep, hamster and mouse scrapie samples.
Proteinase K is useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
It is used for the removal of endotoxins bound to cationic proteins such as lysozyme and ribonuclease A.
It is useful for the isolation of hepatic, yeast, and mung bean mitochondria and is used to determine enzyme localization on membranes
It is used for the treatment of paraffin embedded tissue sections to expose antigen binding sites for antibody labeling and for digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research. Product SAE0009 is provided as a lyophilized powder. Product SAE0009 has been used to break down human lens protein1.
Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.
Reported useful for the isolation of hepatic, yeast, and mung bean mitochondria
Determination of enzyme localization on membranes
Treatment of paraffin embedded tissue sections to expose antigen binding sites for antibody labeling.
Digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research.

Biochem/physiol Actions

Proteinase K has a broad specificity and degrades many proteins even in the native state. It mainly cleaves the peptide bond adjacent to the carboxyl group of aliphatic and aromatic amino acids with blocked a-amino groups.The optimum pH is between 7.5-9.0 and the isoelectric point is 8.9 Ca2+ (1-5 mM) is required for activation. Proteinase K is inhibited by diisopropyl fluorophosphate (DFIP), and phenylmethanesulfonyl fluoride (PMSF).
Proteinase K is a stable and highly reactive serine protease. Evidence from crystal and molecular structure studies indicates the enzyme belongs to the subtilisin family with an active-site catalytic triad (Asp39-His69-Ser224). It is stable in a broad range of environments: pH, buffer salts, detergents (SDS), and temperature. In the presence of 0.1-0.5% SDS, proteinase K retains activity and will digest a variety of proteins and nucleases in DNA preparations without compromising the integrity of the isolated DNA.

Unit Definition

One unit will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 μmole of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of a serine protease proteinase K from Tritirachium album limber at 0.98 ? resolution
Betzel C, et al.
Biochemistry, 40(10), 3080-3088 (2001)
Engineering proteinase K using machine learning and synthetic genes
Liao J, et al.
BMC biotechnology, 7(1), 16-16 (2007)
Proteinase K from Tritirachium album Limber: characterization of the chromosomal gene and expression of the cDNA in Escherichia coli
GUNKEL FA and GASSEN HG
European Journal of Biochemistry, 179(1), 185-194 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service