Skip to Content
Merck
All Photos(1)

Key Documents

SAB5900405

Sigma-Aldrich

Anti-Monkeypox virus/MPXV A29L Antibody (SAA0286)

new

Synonym(s):

A29L, Monkeypox virus (MPXV),A29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

mouse

Quality Level

recombinant

expressed in CHO cells

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

clone

SAA0286, recombinant monoclonal

form

liquid

mol wt

150 kDa

species reactivity

monkeypox virus

concentration

1 mg/mL

technique(s)

ELISA: 1:2000-1:4000
neutralization: 0.01-10 μg/mL

color

colorless

pH

7.4

isotype

IgG2a, kappa

Protein ID accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

-65 to -96°C

target post-translational modification

unmodified

General description

Anti-Monkeypox virus/MPXV A29L Antibody, clone SAA0286, is a monoclonal antibody produced in mouse. It is a single, highly specific, cloned antibody that is directed against the A29L protein of the monkeypox virus. It is composed of two heavy chains and two light chains, linked together by disulfide bonds. The heavy chains contain the antigen-binding region, and the light chains contain the constant region. The antigen-binding region is composed of two variable domains, which are responsible for recognizing and binding to the antigen.


Monkeypox virus (MPXV), like smallpox virus, belongs to the genus Orthopoxvirus. Its morphology is consistent with that of orthopoxvirus, with a brick-shaped or oval shape with rounded corners and a size of 200~400nm. Brick-shaped or oval-shaped, covered by a lipoprotein membrane or envelope, with two protein-containing side bodies in the middle, and a thick membrane core containing a large double-stranded DNA gene body. The genome of Mpox virus is a double-stranded DNA, about 197kb long, and the end of the genome contains an identical but opposite terminal inverted repeat sequence. The virus contains 190 open reading frames, 4 of which are located in terminal inverted repeats. The core proteins of monkeypox virus are A29L, A35R, B6R, M1R, H3L.

Specificity

Monoclonal Anti-Monkeypox virus/MPXV A29L Antibody, clone SAA0286 binds to the A29L protein of the monkeypox virus, thereby inhibiting its ability to replicate and spread. This prevents the virus from causing infection and disease. The antibody also has the ability to neutralize the virus, which further prevents it from causing infection.

Application

ELISA,neutralization

Packaging

100µg per vial / 1 mg per vial

Storage and Stability

Use a manual defrost freezer and avoid repeated freeze-thaw cycles.Store at +4°C short term (1-2 weeks).Store at -20 °C 12 months. Store at -80°C long term.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service