Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
alpaca
Quality Segment
recombinant
expressed in HEK 293 cells (Human cell line)
conjugate
horse radish peroxidase (HRP)
antibody form
affinity purified immunoglobulin
antibody product type
secondary antibodies
clone
recombinant monoclonal
form
lyophilized solid
mol wt
100 kDa
species reactivity
mouse, Alpaca
technique(s)
ELISA: 1:10000-1:50000, western blot: 1:10000-1:50000
size
0.1 mL
color
colorless
isotype
VHH
shipped in
wet ice
storage temp.
2-8°C
General description
Most monoclonal antibodies are generated in mouse. There are five antibody isotypes (IgA, IgD, IgE, IgG, and IgM) from mouse. Each isotype has a different heavy chain. Mouse IgG constitutes 75% of serum immunoglobulins, and IgG is the predominant form of first antibody produced from mouse. Mouse IgG consists of five subclasses-IgG1, IgG2a, IgG2b, IgG2c(inbred mouse strains with the Igh1-b allele have IgG2c isotype instead of IgG2a), IgG3. They are highly homologous and differ mainly in the hinge region. The whole IgG molecule possesses both the Fc region and the Fab region, which possessing the epitope-recognition site. The IgG contains two heavy and light chains, and the heavy chain is about 50 KD and the light chain is about 25 KD. The common IgG is monomeric with a molecular weight of approximately 150 kD.
VHH are single-domain antibodies derived from the variable regions of heavy chain of Camelidae immunoglobulin. The size of VHH is extremely small(less than15KDa) compared to other forms of antibody fragment, which significantly increase the permeability of VHH. Thus VHH is considered of great value for research, diagnostics and therapeutics.
Immunogen
Biochem/physiol Actions
Isotype: VHH domain of alpaca IgG2b/2c fused to goat IgG Fc
Packaging
Physical form
Preparation Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description recombinant Alpaca(VHH) Nano antibody | description recombinant Alpaca(VHH) Nano antibody | description recombinant Alpaca(VHH) Nano antibody | description recombinant Alpaca(VHH) Nano antibody |
| biological source alpaca | biological source alpaca | biological source alpaca | biological source alpaca |
| conjugate horse radish peroxidase (HRP) | conjugate horse radish peroxidase (HRP) | conjugate unconjugated | conjugate unconjugated |
| species reactivity mouse, Alpaca | species reactivity mouse, Alpaca | species reactivity Alpaca, mouse | species reactivity Alpaca, mouse |
| antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin |
| clone recombinant monoclonal | clone recombinant monoclonal | clone recombinant monoclonal | clone recombinant monoclonal |
| form lyophilized solid | form lyophilized solid | form lyophilized solid | form lyophilized solid |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
13 - Non Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



