Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702887

Sigma-Aldrich

Anti-Glycogen synthase 1 (GYS1) Antibody, clone 8N1B3, Rabbit Monoclonal

Synonym(s):

GSY, GYS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
8N1B3, monoclonal
application:
WB
species reactivity:
human, rat, rat
technique(s):
western blot: 1:500 - 1:2000
citations:
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

material

colorless

clone

8N1B3, monoclonal

form

liquid

mol wt

85 kDa

species reactivity

human, rat, rat

concentration

1.05 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

RPASVPPSPSLSRHSSPHQSEDEEDPRNGPLEEDGERYDEDEEAAKDRRNIRAPEWPRRASCTSSTSGSKRNSVDTATSSSLSTPSEPLSPTSSLGEERN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GYS1(2997)

General description

The protein encoded by this gene catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Sep 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 638-737 of human Glycogen synthase (P13807).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service