Skip to Content
Merck
All Photos(2)

Key Documents

SAB5701729

Sigma-Aldrich

Anti-Phospho-c-Jun-S73 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€417.00

€417.00


Estimated to ship on04 May 2025



Select a Size

Change View
100 μL
€417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€417.00


Estimated to ship on04 May 2025


biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

mouse, human, rat

concentration

1.47 mg/mL

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer73)

Gene Information

human ... JUN(3725)

General description

This gene is the putative transforming gene of avian sarcoma virus 17. It encodes a protein which is highly similar to the viral protein, and which interacts directly with specific target DNA sequences to regulate gene expression. This gene is intronless and is mapped to 1p32-p31, a chromosomal region involved in both translocations and deletions in human malignancies.

Immunogen

A synthetic phosphorylated peptide around S73 of human c-Jun (NP_002219.1).

Physical form

Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service