Skip to Content
Merck
All Photos(3)

Key Documents

SAB5700780

Sigma-Aldrich

Anti-NF-kB p65 antibody produced in rabbit

Synonym(s):

CMCU, NF-kB p65, NFKB3, RELA, p65

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€317.00

€317.00


Check Cart for Availability


Select a Size

Change View
100 μL
€317.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€317.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58
59
60

species reactivity

mouse, human, rat

concentration

2.23 mg/ml

technique(s)

immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50-1:100
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RELA(5970)

General description

NF-kappa-B is a ubiquitous transcription factor involved in several biological processes. It is held in the cytoplasm in an inactive state by specific inhibitors. Upon degradation of the inhibitor, NF-kappa-B moves to the nucleus and activates transcription of specific genes. NF-kappa-B is composed of NFKB1 or NFKB2 bound to either REL, RELA, or RELB. The most abundant form of NF-kappa-B is NFKB1 complexed with the product of this gene, RELA. Four transcript variants encoding different isoforms have been found for this gene.

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 50-180 of human NF-kB p65 (NP_068810.3).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service