Skip to Content
Merck
All Photos(8)

Key Documents

SAB5700618

Sigma-Aldrich

Anti-Lamin A/C antibody produced in rabbit

Anti-Lamin A/C antibody produced in rabbit

Synonym(s):

CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, FPLD2, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNA, LMNC, LMNL1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€317.00

€317.00


Check Cart for Availability


Select a Size

Change View
100 μL
€317.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€317.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62-74

species reactivity

human, mouse, rat

concentration

1.41 mg/ml

technique(s)

immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMNA(4000)

General description

The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. Alternative splicing results in multiple transcript variants. Mutations in this gene lead to several diseases: Emery-Dreifuss muscular dystrophy, familial partial lipodystrophy, limb girdle muscular dystrophy, dilated cardiomyopathy, Charcot-Marie-Tooth disease, and Hutchinson-Gilford progeria syndrome.

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 403-572 of human Lamin C (NP_005563.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. Canada
    • Review 1
    • Votes 0
    1 out of 5 stars.

    Performed poorly in Western Blotting

    Purchased this product through Sigma. There was a delay between time of purchase and time of delivery. Product arrived and tested in western blotting at dilution 1:1000 and showed multiple non specific bands on gel making data uninterpretable. Western blotting conditions: Blocking 1hr in 5% Milk TBST, Probed o/night 1:1000 in blocking buffer, after three washes secondary antibody Rabbit HRP at 1:10000 (PTG) and developed with ECL Fempto (UltraScence Femto Western Substrate)

    Helpful?

    1. Response from MilliporeSigma:

      SAB5700618 has been tested for suitability in western blot with a dilution of 1:500-1:2000. Optimal dilution must be determined by the end user. For assistance with determining the optimal dilution for your samples, please contact us for additional technical support, https://www.sigmaaldrich.com/US/en/support/customer-support.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service