Skip to Content
Merck
All Photos(4)

Key Documents

SAB5700095

Sigma-Aldrich

Anti-NDUFB4 antibody produced in rabbit

Synonym(s):

B15, CI-B15, NDUFB4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€317.00

€317.00


Estimated to ship on28 May 2025



Select a Size

Change View
100 μL
€317.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€317.00


Estimated to ship on28 May 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

14
15

species reactivity

human, mouse, rat

concentration

1.23 mg/ml

technique(s)

immunofluorescence: 1:50-1:100
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDUFB4(4710)

General description

This gene encodes a non-catalytic subunit of the multisubunit NADH:ubiquinone oxidoreductase, the first enzyme complex in the mitochondrial electron transport chain (complex I). Mammalian complex I is composed of 45 different subunits and transfers electrons from NADH to ubiquinone.

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human NDUFB4 (NP_001161803.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service