Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600262

Sigma-Aldrich

Anti-TTF1 (Thyroid transcription factor 1) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM373, purified immunoglobulin

Synonym(s):

Homeobox protein Nkx-2.1, Thyroid transcription factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€352.00

€352.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€352.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€352.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM373, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500—1:1000
western blot: 1000-20000

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NKX2-1(7080)

Specificity

This antibody reacts to human TTF1 ((Thyroid transcription factor 1) (Homeobox protein Nkx-2.1). It may also react to mouse and rat TTF1, as predicted by immunogen homology.

Immunogen

A peptide corresponding to N-terminus of human TTF1 ((Thyroid transcription factor 1) (Homeobox protein Nkx-2.1)

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service