Skip to Content
Merck
All Photos(2)

Documents

SAB5600242

Sigma-Aldrich

Anti-CK-5/CK-6 (Cytokeratin 5/ Cytokeratin 6) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM341, purified immunoglobulin

Synonym(s):

Keratin 5, Keratin 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM341, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500—1:1000
western blot: 1:2000-1:4000

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT5(3852)

Specificity

This antibody reacts to both human CK-5 (Cytokeratin 5) and CK-6 (Cytokeratin 6).

Immunogen

A peptide corresponding to the C-terminus of Human Cytokeratin 5

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Fan et al.
Frontiers in oncology, 11, 740410-740410 (2021-12-21)
Craniopharyngiomas (CPs) predominantly involving the third ventricle were commonly termed "intraventricular" lesions. The aim of this study was to clarify the anatomical relationship between the tumor and the third ventricle by both surgical and histological investigation. A retrospective review of
NOTCH4I?L12_16 sensitizes lung adenocarcinomas to EGFR-TKIs through transcriptional down-regulation of HES1.
Zhang, et al.
Nature Communications, 14, 3183-3183 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service