Skip to Content
Merck
All Photos(5)

Key Documents

SAB5600212

Sigma-Aldrich

Anti-Human IgG3 antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM119, purified immunoglobulin

Synonym(s):

IGHG3, Ig gamma 3 chain C, Immunoglobulin gamma 3, Immunoglobulin heavy constant gamma 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€311.00
1 MG
Inquire

€311.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€311.00
1 MG
Inquire

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€311.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM119, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

ELISA: suitable (50ng/well - 200ng/well (for Capture)
0.05ug/mL - 0.2ug/mL (for Detection))

flow cytometry: suitable (assay dependent concentration)
immunocytochemistry: 2-5 μg/mL
immunohistochemistry: 2—5 μg/mL

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

Specificity

This antibody reacts to the heavy chain of human IgG3, RM119 does not cross react to any other IgG subclasses (IgG1, IgG2, or IgG4), and shows no cross reactivity IgM, IgA, IgD, IgE. RM119 does not react to monkey (Cyno or Rhesus) IgG, mouse IgG, rat IgG, or goat IgG.

Immunogen

Human IgG3

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viktoriya B Boncheva et al.
Biomolecules, 12(8) (2022-08-27)
A Crohn's-like lymphoid reaction (CLR) is observed in about 15% of colorectal cancer (CRC) patients and is associated with favourable outcomes. To identify the immune targets recognised by CRC CLR patient sera, we immunoscreened a testes cDNA library with sera

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service