Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600057

Sigma-Aldrich

Anti-p38 MAPK Phospho (pT180/pY182) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM243, purified immunoglobulin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€367.00

€367.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€367.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€367.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM243, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 1:1000-1:2000
immunohistochemistry: 1:500-1:1000

isotype

IgG

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr180/pTyr182)

Gene Information

human ... MAPK14(1432)

General description

The mitogen activated protein kinase (MAPK) gene codes for a serine/threonine protein kinase which is found to be expressed in majority of cells. The MAPK gene is mapped to human chromosome 6p21.31 and the encoded protein consists of 90 to 360 amino acids residues. MAPKs are divided into three subtypes : extracellular signal-regulated kinases (ERK), c-Jun amino-terminal kinases (JNK) and p38 MAPK (p38).

Specificity

This antibody reacts to p38 MAPK only when dual phosphorylation at Thr180 and Tyr182 is observed. There is no cross-reactivity with p38 MAPK without dual phosphorylation at Thr180 and Tyr182.

Immunogen

Synthetic phospho-peptide corresponding to human Phospho-p38 MAPK (Thr180/Tyr182)

Application

Anti-p38 MAPK Phospho (pT180/pY182) antibody, Rabbit monoclonal has been used for immunodetection.

Biochem/physiol Actions

Mitogen activated protein kinase (MAPK) participates in transdifferentiation of adipocytes in myoblast by regulating adipogenic and adipolysis metabolism. p38 MAPK is associated with various biological processes as such as cell proliferation and differentiation, cell adhesion, migration. It mediates pro-apoptotic signal during apoptosis, contributing either to cell death or survival.. p38 MAPK is known to mediate a number of metabolic pathways by controlling transcription, mRNA stability, chromatin remodelling and protein synthesis. During initial process of tumorigenesis, It acts as a tumor suppressor, while it might promote metastasis in the later stages of the same. p38 MAPK gets activated in response to growth factors, oxidative stress, proinflammatory cytokines and DNA damage.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MAPK14 (mitogen-activated protein kinase 14).
Porras A and Guerrero Arroyo M D C
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2011)
Expressions and regulatory effects of P38/ERK/JNK MAPKs in the adipogenic trans-differentiation of C2C12 myoblasts.
Qi R, et al.
Cellular Physiology and Biochemistry, 44(6), 2467-2475 (2017)
TINCR facilitates non-small cell lung cancer progression through BRAF-activated MAPK pathway.
Zhu Z J and He J K
Biochemical and Biophysical Research Communications, 497(4), 971-977 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service