Skip to Content
Merck
All Photos(2)

Key Documents

SAB56000338

Sigma-Aldrich

Anti-PAX8 Rabbit Monoclonal Antibody

clone RM436, affinity purified immunoglobulin

Synonym(s):

PAX8, Paired box protein Pax-8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€370.00

€370.00


Check Cart for Availability


Select a Size

Change View
100 μL
€370.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€370.00


Check Cart for Availability

antibody form

affinity purified immunoglobulin

Quality Level

antibody product type

primary antibodies

clone

RM436, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: 1:100-1:400

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAX8(7849)

Specificity

This antibody reacts to human Paired box protein Pax-8. It may also react to mouse and rat Paired box protein Pax-8, as predicted by immunogen homology.

Immunogen

A peptide corresponding to corresponding to C-terminus of human Paired box protein Pax-8

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service