Skip to Content
Merck
All Photos(3)

Key Documents

SAB5500102

Sigma-Aldrich

Anti-EZH2 antibody, Rabbit monoclonal

clone SP129, recombinant, expressed in proprietary host, affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€338.00

€338.00


Check Cart for Availability


Select a Size

Change View
100 μL
€338.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€338.00


Check Cart for Availability

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP129, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

monkey, frog, mouse, rat

technique(s)

flow cytometry: 1:100
immunoblotting: 1:25
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EZH2(2146)

General description

KMT6/Enhancer of zeste homologue 2 (EZH2) is a histone lysine methyl transferase associated with transcriptional repression. It is a component of the polycomb repressive complexes PRC2 and PRC3. The PRC2 complex catalyses histone H3 K27 trimethylation. EZH2 is ubiquitously expressed during early embryo genesis, and becomes restricted to the central and peripheral nervous systems and sites of fetal hematopoiesis during later development. EZH2 is highly expressed in follicular T-cells and in a variety of tumors such as lymphoma, breast and prostate.

Immunogen

Synthetic peptide corresponding to C-terminus of human EZH2 protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivan Osokine et al.
Cell reports, 38(5), 110329-110329 (2022-02-03)
A little-appreciated feature of early pregnancy is that embryo implantation and placental outgrowth do not evoke wound-healing responses in the decidua, the specialized endometrial tissue that surrounds the conceptus. Here, we provide evidence that this phenomenon is partly due to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service