Skip to Content
Merck
All Photos(2)

Key Documents

SAB5500022

Sigma-Aldrich

Anti-E-Cadherin antibody, Rabbit monoclonal

clone SP64, recombinant, expressed in proprietary host, affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€338.00

€338.00


Check Cart for Availability


Select a Size

Change View
100 μL
€338.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€338.00


Check Cart for Availability

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP64, monoclonal

species reactivity

human (tested)

technique(s)

immunoblotting: 1:25
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CDH1(999)

General description

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium dependent cell adhesion molecule expressed predominately in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

Immunogen

Recombinant protein encoding aa 600-707 of human cadherin-E.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Lacerda Coelho et al.
Biology, 13(7) (2024-07-26)
Breast cancer is the most prevalent cancer among women worldwide. Therapeutic strategies to control tumors and metastasis are still challenging. Three-dimensional (3D) spheroid-type systems more accurately replicate the features of tumors in vivo, working as a better platform for performing
Cheng Zhang et al.
Oncology letters, 13(4), 2059-2070 (2017-04-30)
Human bone marrow stromal cells (hBMSCs) may contribute to the growth of tyrosine kinase inhibitor (TKI)-resistant chronic myelogenous leukemia (CML). However, there are certain differences in biology between CML and Philadelphia-positive acute lymphoblastic leukemia (Ph+ ALL). Little is known about
Luis G V Fernandes et al.
Frontiers in microbiology, 14, 1199660-1199660 (2023-07-10)
Leptospirosis is a worldwide zoonosis caused by pathogenic and virulent species of the genus Leptospira, whose pathophysiology and virulence factors remain widely unexplored. Recently, the application of CRISPR interference (CRISPRi) has allowed the specific and rapid gene silencing of major
Juan Zhou et al.
Oncology reports, 39(4), 1783-1792 (2018-02-03)
Chemotherapy is one of the methods to treat patients with non-small cell lung cancer (NSCLC) developing resistance to epidermal growth factor receptor (EGFR) tyrosine kinase inhibitors (TKIs), such as gefitinib. Previous studies revealed that the sensitivity to chemotherapy may depend

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service