Skip to Content
Merck
All Photos(2)

Key Documents

SAB5202340

Sigma-Aldrich

Monoclonal Anti-O-GalNAC-FITC antibody produced in mouse

clone 9B9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€278.00

€278.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€278.00

About This Item

UNSPSC Code:
12352203

€278.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9B9, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1.0 mg/mL

technique(s)

immunofluorescence: 1:50
western blot: 1:1000

isotype

IgM

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

glycosylation

Immunogen

Synthetic β-O-Linked N-Acetylglucosamine conjugated to Keyhole Limpet Hemocyanin (KLH).

Physical form

PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service