Skip to Content
Merck
All Photos(3)

Key Documents

SAB5201403

Sigma-Aldrich

Monoclonal Anti-Mitofusin - Biotin antibody produced in mouse

clone S153-5, purified immunoglobulin

Synonym(s):

Anti-CMT2A, Anti-CPRP 1,, Anti-KIAA0214, Anti-Mitochondrial assembly regulatory factor, Anti-hyperplasia suppressor gene

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€568.00

€568.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€568.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€568.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S153-5, monoclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt 90 kDa

species reactivity

rat, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Mfn2(170731)

Immunogen

Fusion protein amino acids 370-600 (cytoplasmic N-terminus) of mouse Mitofusin-2. Rat: 97% identity (226/231 amino acids identical). Human: 92% identity (214/231 amino acids identical) ~55% identity with Mitofusin-1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS pH 7.4, 50% glycerol, 0.1% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service