Select a Size
About This Item
Skip To
biological source
mouse
Quality Level
conjugate
Atto 390 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
S289-16, monoclonal
form
buffered aqueous solution
mol wt
antigen predicted mol wt 160 kDa
species reactivity
rat, hamster, mouse
concentration
1 mg/mL
technique(s)
immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
rat ... Abcc8(25559)
1 of 4
This Item | |||
|---|---|---|---|
| conjugate Atto 390 conjugate | conjugate Atto 488 conjugate | conjugate Atto 594 conjugate | conjugate unconjugated |
| antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| Gene Information rat ... Abcc8(25559) | Gene Information rat ... Abcc8(25559) | Gene Information rat ... Abcc8(25559) | Gene Information rat ... Abcc8(25559) |
| biological source mouse | biological source mouse | biological source mouse | biological source mouse |
| species reactivity rat, hamster, mouse | species reactivity mouse, rat, hamster | species reactivity rat, mouse, hamster | species reactivity rat, mouse, hamster |
| clone S289-16, monoclonal | clone S289-16, monoclonal | clone S289-16, monoclonal | clone S289-16, monoclonal |
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SAB5200529-100UG | 04061832147291 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service