Skip to Content
Merck
All Photos(2)

Key Documents

SAB5200107

Sigma-Aldrich

Anti-SCNN1G antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-ENaC Gamma, Anti-Scnn1g

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€371.00

€371.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€371.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€371.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 83 kDa

species reactivity

rat, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Sodium channel epithelial 1 γ subunit (SCNN1G) is one of the subunits of epithelial sodium channels. The gene ID of the protein is 24768.

Specificity

Detects ~83 kDa.

Immunogen

AA629–650 of the rat sequence

Application

Anti-SCNN1G antibody produced in rabbit has been used for Western blotting.

Biochem/physiol Actions

Sodium channel epithelial 1 γ subunit (SCNN1G) is involved in sodium transport in the epithelium.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zaixing Chen et al.
American journal of physiology. Lung cellular and molecular physiology, 307(8), L609-L617 (2014-08-31)
Epithelial sodium channels (ENaC) govern transepithelial salt and fluid homeostasis. ENaC contributes to polarization, apoptosis, epithelial-mesenchymal transformation, etc. Fibrinolytic proteases play a crucial role in virtually all of these processes and are elaborated by the airway epithelium. We hypothesized that
Adverse effect of assisted reproductive technology-related hyperoestrogensim on the secretion and absorption of uterine fluid in superovulating mice during the peri-implantation period.
Xia, et al.
Frontiers in Endocrinology, 14, 859204-859204 (2023)
Md Shahrier Amin et al.
Hypertension (Dallas, Tex. : 1979), 54(4), 860-867 (2009-07-29)
To elucidate the role of epithelial sodium channels (ENaCs) and Na(+)-K(+)-ATPase in Na(+) transport by the choroid plexus, we studied ENaC expression and Na(+) transport in the choroid plexus. Lateral ventricle choroid plexuses were obtained from young male Wistar, Dahl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service