Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200103

Sigma-Aldrich

Anti-SLC12A1 antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-NKCC2, Anti-Slc12a1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€371.00

€371.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€371.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€371.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 160 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 12 member 1 (SLC12A1) is a Na+-K+-Cl- transporter and it is expressed in the thick ascending loop of kidneys in rats. The gene ID of the protein is 25065.

Specificity

Detects ~160 kDa.

Immunogen

AA109 to 129 of the rat sequence

Biochem/physiol Actions

Solute carrier family 12 member 1 (SLC12A1) functions in water reabsorption in the kidneys.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuko Itoh et al.
Biochemical and biophysical research communications, 453(3), 356-361 (2014-09-30)
Sodium reabsorption via Na-K-2Cl cotransporter 2 (NKCC2) in the thick ascending limbs has a major role for medullary osmotic gradient and subsequent water reabsorption in the collecting ducts. We investigated intrarenal localization of three isoforms of NKCC2 mRNA expressions and
Yanting Chen et al.
Frontiers in physiology, 12, 642274-642274 (2021-04-20)
Emerging evidence is showing that apelin plays an important role in regulating salt and water balance by counteracting the antidiuretic action of vasopressin (AVP). However, the underlying mechanism remains unknown. Here, we hypothesized that (pro) renin receptor (PRR)/soluble prorenin receptor
Gustavo R Ares et al.
Hypertension (Dallas, Tex. : 1979), 60(6), 1464-1470 (2012-10-31)
Salt-sensitive hypertension involves a renal defect preventing the kidney from eliminating excess NaCl. The thick ascending limb of Henle loop reabsorbs ≈ 30% of filtered NaCl via the apical Na-K-2Cl cotransporter (NKCC2). Higher NKCC2 activity and Cl reabsorption have been
Aurélie Edwards et al.
American journal of physiology. Renal physiology, 307(2), F137-F146 (2014-05-23)
This study aims to understand the extent to which modulation of the Na(+)-K(+)-2Cl(-) cotransporter NKCC2 differential splicing affects NaCl delivery to the macula densa. NaCl absorption by the thick ascending limb and macula densa cells is mediated by apical NKCC2.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service