Skip to Content
Merck
All Photos(2)

Key Documents

SAB5200101

Sigma-Aldrich

Anti-BSN antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti- Neuronal double zine finger protein, Anti-BSN, Anti-Bassoon, Anti-ZNF231

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€524.00

€524.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1779


Select a Size

Change View
100 μG
€524.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€524.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1779

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 420 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BSN(8927)

General description

Bassoon presynaptic cytomatrix protein (BSN) is a 420kDa protein which is expressed in the neuronal synapses. The gene encoding it is located on human chromosome 3p21. The gene ID of the protein is 8927.

Specificity

Detects ~420 kDa, corresponding to the molecular mass of Bassoon. Multiple isoforms can be detected.

Immunogen

NM_003458.3 (AA 786-1041) N-terminal his-tagged fusion protein

Biochem/physiol Actions

Bassoon presynaptic cytomatrix protein (BSN) acts as a scaffold protein and mediates release of neurotransmitters. It is also a part of trafficking and helps in delivery of proteins across the synapse.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, pH 7..4, 50% glycerol, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Winter et al.
Genomics, 57(3), 389-397 (1999-05-18)
Bassoon is a novel 420-kDa protein recently identified as a component of the cytoskeleton at presynaptic neurotransmitter release sites. Analysis of the rat and mouse sequences revealed a polyglutamine stretch in the C-terminal part of the protein. Since it is
Stefan Hallermann et al.
Neuron, 68(4), 710-723 (2010-11-26)
Sustained rate-coded signals encode many types of sensory modalities. Some sensory synapses possess specialized ribbon structures, which tether vesicles, to enable high-frequency signaling. However, central synapses lack these structures, yet some can maintain signaling over a wide bandwidth. To analyze
Nature, 447(7145), 661-678 (2007-06-08)
There is increasing evidence that genome-wide association (GWA) studies represent a powerful approach to the identification of genes involved in common human diseases. We describe a joint GWA study (using the Affymetrix GeneChip 500K Mapping Array Set) undertaken in the
Christopher Patzke et al.
Cell, 179(2), 498-513 (2019-10-05)
Neuromodulators bind to pre- and postsynaptic G protein-coupled receptors (GPCRs), are able to quickly change intracellular cyclic AMP (cAMP) and Ca2+ levels, and are thought to play important roles in neuropsychiatric and neurodegenerative diseases. Here, we discovered in human neurons

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service