Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200099

Sigma-Aldrich

Anti-HSPB2 antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Anti-HSPB2 (MKBP), Anti-DMPK-binding protein, Anti-HspB2, Anti-MGC133245, Anti-heat shock 27kDa protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€371.00

€371.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€371.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€371.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 27 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPB2(3316)

General description

Heat shock protein family B, member 2 (HSPB2) is also called as myotonic dystrophy kinase binding protein (MKBP). It is expressed in the skeletal muscles and heart. Studies have also shown that HSPB2 is overexpressed in lung fibrosis. The gene encoding it is localized on human chromosome 11q22-q23. The gene ID of the protein is 3316.

Specificity

Detects ~27 kDa.

Immunogen

Synthetic peptide

Biochem/physiol Actions

Heat shock protein family B, member 2 (HSPB2) binds to ubiquitin and hydrogen peroxide-inducible clone 5, an adaptor protein. This association is important for the ubiquitination and degradation of NADPH oxidase 4 (Nox4). It is also crucial for the degradation of other proteins. HSPB2 interacts with myotonic dystrophy protein kinase (DMPK) and activates it.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, 50% glycerol, and 0.02% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leena P Desai et al.
The Journal of biological chemistry, 289(26), 18270-18278 (2014-05-17)
Hydrogen peroxide-inducible clone 5 (Hic-5) is a focal adhesion adaptor protein induced by the profibrotic cytokine TGF-β1. We have demonstrated previously that TGF-β1 induces myofibroblast differentiation and lung fibrosis by activation of the reactive oxygen species-generating enzyme NADPH oxidase 4
Xiaofei Chang et al.
International journal of oncology, 38(4), 1129-1135 (2011-01-25)
Hypermethylation of gene promoters and the corresponding loss of gene expression are recognized as a hallmark of human cancer, and DNA methylation has emerged as a promising biomarker for the detection of human esophageal squamous cell carcinoma (ESCC). To identify
Sankaralingam Prabhu et al.
PloS one, 7(1), e29810-e29810 (2012-01-25)
The small heat shock protein, human HspB2, also known as Myotonic Dystrophy Kinase Binding Protein (MKBP), specifically associates with and activates Myotonic Dystrophy Protein Kinase (DMPK), a serine/threonine protein kinase that plays an important role in maintaining muscle structure and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service