Skip to Content
Merck
All Photos(1)

Documents

SAB5200063

Sigma-Aldrich

Monoclonal Anti-KCNA1 antibody produced in mouse

clone S36-15, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-KCNA2B, Anti-Kv1.1, Anti-Kv1.1 Extracellular, S36-15, Anti-kcna1, Anti-kcpvd

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S36-15, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 56 kDa

species reactivity

mouse, rat

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Specificity

Detects ~56 kDa (could be 65-85 depending on glycosylation)

Immunogen

Synthetic peptide amino acids 191-208 (ELKDDKDFTGTIHRIDNTC, extracellular domain) or rat Kv1.1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service