Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200055

Sigma-Aldrich

Monoclonal Anti-KCNB1 antibody produced in mouse

clone S4-11, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-DRK1PC, Anti-Kcnb1, Anti-Kcr1-1, Anti-Kv2.1, Anti-Kv2.1, S4-11, Anti-Shab

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€420.00

€420.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€420.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€420.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S4-11, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 105-125 kDa

species reactivity

human, mouse, rat, monkey

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The potassium voltage-gated channel subfamily B member 1 (KCNB1) gene is mapped to human chromosome 20q13.13. This gene codes for a α subunit of voltage-gated potassium Kv2.1 channel. The encoded protein contains 858 amino acids and six transmembrane segments (S1–S6). KCNB1 is mainly expressed in the central nervous system, but at high level in somatodendritic compartment of neocortical and pyramidal neurons.

Specificity

Detects ~105-125 kDa. Recognizes the Shab related α-subunit Kv2.1.

Immunogen

Bacterially expressed GST Bacterially expressed GSTfusion protein corresponding toamino acids 509-853 of rat KV2.1, Accession Number NM_004975.

Biochem/physiol Actions

Potassium voltage-gated channel subfamily B member 1 (KCNB1) plays a vital role in hippocampal neuron excitation homeostasis. The encoded protein is mainly implicated in the generation of the major delayed rectifier potassium current in pyramidal neurons. Mutation in the gene is associated with the development of left ventricular (LV) hypertrophy and early-onset infantile epilepsy in humans.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De novo KCNB1 mutations in infantile epilepsy inhibit repetitive neuronal firing
Saitsu H
Scientific Reports (2015)
Novel KCNB1 mutation associated with non-syndromic intellectual disability
Latypova X
Journal of Human Genetics, 62, 569-573 (2017)
Genome-wide association study identifies single-nucleotide polymorphism in KCNB1 associated with left ventricular mass in humans: The HyperGEN Study
Arnett DK
BMC Medical Genetics (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service