Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700773

Sigma-Aldrich

Monoclonal Anti-Sialyl Lewis a antibody produced in mouse

clone 121SLE, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€224.00

€224.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€224.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€224.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

121SLE, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable

isotype

IgM

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

The antibody 121SLE recognizes Sialyl Lewis a containing glycolipids.

Immunogen

Immunoprecipitate obtained after immunodiffusion of MAb 19-9 and mucins isolated from an ovarian cyst of a 0Le(a+b-) patient.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Kolben et al.
Journal of cancer research and clinical oncology, 148(12), 3323-3335 (2022-06-22)
Endometrial cancer (EC) is the most common gynecological cancer worldwide. Treatment has been improved in recent years, but, in advanced stages, therapeutical options are still limited. It has been reported that the expression of the blood group antigens Sialyl Lewis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service