Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700699

Sigma-Aldrich

IgM-FITC Isotype Control murine

clone PFR-03, purified immunoglobulin, buffered aqueous solution

Synonym(s):

IgM-Fluorescein isothiocyanate Isotype Control murine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.42
Pricing and availability is not currently available.

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

clone

PFR-03, monoclonal

form

buffered aqueous solution

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgM

shipped in

wet ice

storage temp.

2-8°C

General description

This mouse IgM monoclonal antibody (clone PFR-03) reacts with undefined epitope on a plant pathogen.

Immunogen

Phytophthora sp.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in Tris buffered saline, pH 8.0, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service