Skip to Content
Merck
All Photos(1)

Documents

SAB4700154

Sigma-Aldrich

Monoclonal Anti-CD33-Biotin antibody produced in mouse

clone HIM3-4, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HIM3-4, monoclonal

form

buffered aqueous solution

species reactivity

human, nonhuman primates

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD33(945)

General description

Myeloid cell surface antigen CD33, (cluster of differentiation 33) also known as SIGLEC3, siglecs (sialic acid binding Ig-like lectins) and GP67, belongs to the CD33-related SIGLEC gene family. It is a type 1 transmembrane protein and contains two immunoglobulin-like extracellular domains, a single transmembrane area and two intracellular inhibitory motifs. The CD33 gene is mapped to human chromosome 19q13.41.
The antibody HIM3-4 reacts with CD33, a 67 kDa type I transmembrane glycoprotein (immunoglobulin superfamily) expressed on myeloid progenitors, monocytes, granulocytes, dendritic cells and mast cells; it is absent on platelets, lymphocytes, erythrocytes and hematopoietic stem cells.

Immunogen

NFMY-9s human cell line

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1:1000. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Association of CD33 (cluster of differentiation 33) with sialic acid-containing ligands causes suppression of proinflammatory pathways in monocytes and macrophages. It is mainly involved in anti-inflammatory signaling, cell adhesion and endocytosis functions. CD33 also works as a myeloid differentiation marker. Mutations in the CD33 gene are associated with Alzheimer′s disease susceptibility and acute myeloid leukemia treatment efficacy. In microglia, increase in the expression of CD33 decreases amyloid β peptide

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The "Alzheimer's disease signature": potential perspectives for novel biomarkers
Davinelli S
Immunity & ageing : I & A (2011)
CD33 rs3865444 Polymorphism Contributes to Alzheimer's Disease Susceptibility in Chinese, European, and North American Populations.
Li X, et al.
Molecular Neurobiology, 52, 414-414 (2015)
Human-specific derived alleles of CD33 and other genes protect against postreproductive cognitive decline.
Schwarz F, et al.
Proceedings of the National Academy of Sciences of the USA, 113, 74-74 (2016)
Association of CD33 polymorphism rs3865444 with Alzheimer's disease pathology and CD33 expression in human cerebral cortex.
Walker DG, et al.
Neurobiology of Aging, 36, 571-571 (2015)
Manasi Malik et al.
Human molecular genetics, 24(12), 3557-3570 (2015-03-13)
The CD33 single-nucleotide polymorphism (SNP) rs3865444 has been associated with the risk of Alzheimer's disease (AD). Rs3865444 is in linkage disequilibrium with rs12459419 which has been associated with efficacy of an acute myeloid leukemia (AML) chemotherapeutic agent based on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service