SAB4600158
Anti-Chicken IgY (H+L), highly cross-adsorbed, CF™ 640R antibody produced in goat
~2 mg/mL, affinity isolated antibody
Select a Size
€74.10
Select a Size
About This Item
€74.10
biological source
goat
conjugate
CF™ 640R conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
chicken
concentration
~2 mg/mL
technique(s)
flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect immunofluorescence: 1-10 μg/mL
fluorescence
λex 642 nm; λem 662 nm
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Related Categories
Specificity
Immunogen
Features and Benefits
Physical form
Preparation Note
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service