Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504640

Sigma-Aldrich

Anti-phospho-PLK1 (pSer137) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer137)

Gene Information

human ... PLK1(5347)

Related Categories

General description

Polo-like kinase 1 (PLK1) belongs to the polo-like kinase family. PLK1 is characterized with two COOH-terminal polo box domain and N-terminal kinase domain involved in drug development. PLK1 is expressed primarily at centrosomes and at the midbody of dividing cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human PLK1 around the phosphorylation site of Ser137.

Immunogen Range: 103-152

Biochem/physiol Actions

Polo-like kinase 1 (PLK1) is involved in regulation of mitosis. PLK1 plays a crucial role in centrosome-mediated microtubule events and spindle assembly. PLK1 expression is elevated in cancer tissues. Thus, it can be used as a potential prognostic marker for several cancers, including non-small-cell lung cancer, squamous-cell carcinomas of the head and neck, melanomas, colorectal cancer, oropharyngeal carcinomas, and ovarian and endometrial carcinomas. PLK1 recruited to centrosomes during S-phase by centrosomal protein FOP-related protein of 20kDa (FOR20), plays a vital role in S-phase progression. The protein also plays an essential role in central spindle formation and cytokinesis. PLK1 inhibitors might act as a potential therapeutics in cancer treatment.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minhong Shen et al.
Cell research, 23(11), 1284-1295 (2013-09-11)
Centrosomes are required for efficient cell cycle progression mainly by orchestrating microtubule dynamics and facilitating G1/S and G2/M transitions. However, the role of centrosomes in S-phase progression is largely unknown. Here, we report that depletion of FOR20 (FOP-related protein of
Klaus Strebhardt et al.
Nature reviews. Cancer, 6(4), 321-330 (2006-03-25)
Human polo-like kinase 1 (PLK1) is essential during mitosis and in the maintenance of genomic stability. PLK1 is overexpressed in human tumours and has prognostic potential in cancer, indicating its involvement in carcinogenesis and its potential as a therapeutic target.
Takao Takahashi et al.
Cancer science, 94(2), 148-152 (2003-04-24)
PLK (polo-like kinase), the human counterpart of polo in Drosophila melanogaster and of CDC5 in Saccharomyces cerevisiae, belongs to a family of serine/threonine kinases. It is intimately involved in spindle formation and chromosome segregation during mitosis. The purpose of this
Birgit Spänkuch-Schmitt et al.
Journal of the National Cancer Institute, 94(24), 1863-1877 (2002-12-19)
Expression of polo-like kinase-1 (PLK1), which has several functions in mitotic progression, is elevated in a broad range of human tumors. To investigate the role of PLK1 in neoplastic proliferation, we used the technique of RNA interference. Cells from several
K E Mundt et al.
Biochemical and biophysical research communications, 239(2), 377-385 (1997-11-05)
The human protein kinase Plk1, a member of the polo-like kinase family, is known to function at mitosis. Here we show that the relative specific activity of Plk1 increases in mitosis, that Plk1 is specifically phosphorylated during mitosis, and that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service