Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504621

Sigma-Aldrich

Anti-phospho-MER/SKY (pTyr749/681) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€462.00

€462.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€462.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€462.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 110 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr749/pTyr681)

Gene Information

General description

Tyro3 (TYRO3 protein tyrosine kinase) is an 890 amino acid protein that belongs to the TAM (Tyro3, Axl and Mer) receptor family. Tyro3 is mapped to human chromosome 15q15. It is highly expressed in the nervous system like the neocortex, hippocampus and cerebellum. It is also present in monocytes, macrophages, platelets, dendritic cells and NK cells, breast, ovary, testis, lung, kidney, retinal pigment epithelium and osteoclasts.

Immunogen

The antiserum was produced against synthesized peptide derived from human MER/SKY around the phosphorylation site of Tyr749/681.

Immunogen Range: 716-765

Biochem/physiol Actions

Tyro3 (TYRO3 protein tyrosine kinase) serves as a therapeutic target in breast cancer. This protein is involved in spermatogenesis, immunoregulation, and phagocytosis. It controls disseminated prostate cancer cellular dormancy. TYRO3 helps in the survival and transport of of gonadotropin-releasing hormone (GnRH) neurons within the forebrain. Mutation in TYRO3 result in retinitis pigmentosa.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TYRO3 as a potential therapeutic target in breast cancer.
Ekyalongo RC, et al.
Anticancer Research, 34(7), 3337-3345 (2014)
Nour Al Kafri et al.
Biochemistry and biophysics reports, 30, 101263-101263 (2022-05-07)
The homologous proteins Gas6 and protein S (ProS1) are both natural ligands for the TAM (Tyro3, Axl, MerTK) receptor tyrosine kinases. ProS1 selectively activates Tyro3; however, the precise molecular interface of the ProS1-Tyro3 contact has not been characterised. We used
Mer tyrosine kinase regulates disseminated prostate cancer cellular dormancy.
Cackowski FC, et al.
Journal of Cellular Biochemistry, 118(4), 891-902 (2017)
Ligand Recognition and Homophilic Interactions in Tyro3 STRUCTURAL INSIGHTS INTO THE Axl/Tyro3 RECEPTOR TYROSINE KINASE FAMILY.
Heiring C, et al.
The Journal of Biological Chemistry, 279(8), 6952-6958 (2004)
A novel MERTK mutation causing retinitis pigmentosa.
Al-Khersan H, et al.
Graefe'S Archive For Clinical and Experimental Ophthalmology = Albrecht Von Graefes Archiv Fur Klinische Und Experimentelle Ophthalmologie, 255(8), 1613-1619 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service