Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504315

Sigma-Aldrich

Anti-phospho-PI3-kinase p85-α (pTyr607) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AGM7, Anti-GRB1, Anti-IMD36, Anti-p85, Anti-p85-ALPHA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€462.00

€462.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€462.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€462.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 83 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr607)

Gene Information

human ... PIK3R1(5295)

Related Categories

General description

PIK3R1 (phosphoinositide-3-kinase regulatory subunit 1) is a pleiotropic gene, that codes for a p85α regulatory subunit of phosphatidylinositol-3 kinase (PI3K). This gene is mapped to human chromosome 5q13.

Immunogen

The antiserum was produced against synthesized peptide derived from human PI3-kinase p85-alpha around the phosphorylation site of Tyr607.

Immunogen Range: 573-622

Biochem/physiol Actions

PIK3R1 (phosphoinositide-3-kinase regulatory subunit 1) is known to mediate immune cell differentiation, development and function. Mutations in PIK3R1 gene result in SHORT syndrome, hyper IgM syndrome, lymphadenopathy and short stature.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amplification of telomerase (hTERT) gene is a poor prognostic marker in non-small-cell lung cancer.
Zhu C Q, et al.
British Journal of Cancer, 94(10), 1452-1452 (2006)
Dominant Splice Site Mutations in PIK3R1 Cause Hyper IgM Syndrome, Lymphadenopathy and Short Stature.
Petrovski S
Journal of clinical immunology, 36(5), 462-471 (2016)
Iris Malformation and Anterior Segment Dysgenesis in Mice and Humans With a Mutation in PI 3-Kinase.
Solheim MH
Investigative Ophthalmology & Visual Science, 58(7), 3100-3106 (2017)
Qiaoni Hu et al.
Journal of biochemical and molecular toxicology, 37(1), e23220-e23220 (2022-09-13)
We have identified that hydrogen sulfide (H2 S), a gaseous mediator, plays a crucial role in antioxidative, anti-inflammatory, and cytoprotective effects on uranium (U)-triggered rat nephrotoxicity. Pyroptosis is a special mode of inflammation and programmed cell death involved in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service