Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503940

Sigma-Aldrich

Anti-phospho-MITF (pSer180/73) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CMM8, Anti-COMMAD, Anti-MI, Anti-MITF-A, Anti-WS2, Anti-WS2A, Anti-bHLHe32

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€462.00

€462.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€462.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€462.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 58 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer180/pSer73)

Gene Information

human ... MITF(4286)

Immunogen

The antiserum was produced against synthesized peptide derived from human MITF around the phosphorylation site of Ser180/73.

Immunogen Range: 151-200

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Suzuki et al.
EMBO reports, 24(7), e56574-e56574 (2023-05-22)
Dysregulation of the activity of the mechanistic target of rapamycin complex 1 (mTORC1) is commonly linked to aging, cancer, and genetic disorders such as tuberous sclerosis (TS), a rare neurodevelopmental multisystemic disease characterized by benign tumors, seizures, and intellectual disability.
Nunzia Caporarello et al.
International journal of molecular medicine, 40(4), 1277-1284 (2017-08-30)
The mechanisms underlying cutaneous melanogenesis have been widely studied; however, very little is known about uveal melanogenesis. Melanin is normally produced by uveal melanocytes and gives the color to the iris. A derangement from this normal production may occur, for
Linxin Li et al.
Nature communications, 9(1), 2685-2685 (2018-07-12)
How cells coordinate the response to fluctuating carbon and nitrogen availability required to maintain effective homeostasis is a key issue. Amino acid limitation that inactivates mTORC1 promotes de-phosphorylation and nuclear translocation of Transcription Factor EB (TFEB), a key transcriptional regulator
Lele Li et al.
Journal of cellular and molecular medicine, 27(10), 1327-1340 (2023-04-19)
Choroidal neovascularization (CNV), is a major cause of irreversible blindness among the elderly population in developed countries, which is resulted from subretinal fibrosis without effective therapeutic strategies. Endothelial-to-mesenchymal transition (EndMT) of choroidal vascular endothelial cells (CVECs) contributes to subretinal fibrosis.
Claire Regazzetti et al.
The Journal of investigative dermatology, 138(1), 171-178 (2017-08-27)
The shorter wavelengths of the visible light spectrum have been recently reported to induce a long-lasting hyperpigmentation but only in melano-competent individuals. Here, we provide evidence showing that OPN3 is the key sensor in melanocytes responsible for hyperpigmentation induced by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service