Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503582

Sigma-Aldrich

Anti-KCNAB3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

K(+) channel subunit β-3, KCNA3B, KCNAB3, Kv-β-3, Voltage-gated potassium channel subunit β-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNAB3(9196)

General description

The gene coding for potassium voltage-gated channel subfamily A regulatory β subunit 3 (KCNAB3) is mapped to human chromosome 17p13.1. Human Kvβ3 transcript is mainly expressed in the brain, with high expression levels in cerebellum.

Immunogen

The antiserum was produced against synthesized peptide derived from human KCNAB3.

Immunogen Range: 293-342

Biochem/physiol Actions

Potassium voltage-gated channel subfamily A regulatory β subunit 3 (KCNAB3) is involved in migraine pathogenesis.2 Kvβ3 expressed along with Kv1.5 in Chinese hamster ovary cells gives rise to a new A-type potassium channel.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coexpression of the KCNA3B gene product with Kv1.5 leads to a novel A-type potassium channel.
Leicher T
The Journal of Biological Chemistry, 273(52), 35095-35101 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service