Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503568

Sigma-Aldrich

Anti-EXO1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

EXO1, Exonuclease 1, Exonuclease I, hExo1, hExoI

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 94 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EXO1(9156)

General description

Anti-EXO1 Antibody detects endogenous levels of total EXO1 protein.
Exonuclease 1 (EXO1), a member of RAD2 nuclease family, comprises an amino-terminal (N) domain, intermediate spacer region and an internal (I) domain with cysteine and glutamate residues. The Exo 1 gene is mapped to human chromosome 1q43.

Immunogen

The antiserum was produced against synthesized peptide derived from human EXO1.

Immunogen Range: 61-110

Application

Anti-EXO1 antibody produced in rabbit has been used in immunoblotting.[1]

Biochem/physiol Actions

Exonuclease 1 (EXO1) N-domain mediates the DNA binding and the I domain is crucial for magnesium binding. It displays micro-mediated end-joining, 5′ to 3′ exonuclease activity and mediates homologous recombination and replication. EXO1 is more efficient on single-stranded DNA (ssDNA) than double-stranded DNA in exonucleolytic degradation. It also exhibits 5′ ssDNA-flap-specific endonuclease activity. EXO1 polymorphisms may be implicated in epithelial ovarian cancer (EOC).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingyan Shi et al.
OncoTargets and therapy, 10, 4841-4851 (2017-10-19)
Exonuclease 1 (EXO1), one of DNA mismatch repair pathway genes, functions in maintaining genomic stability and affects tumor progression. We hypothesized that genetic variations in EXO1 may predict clinical outcomes in epithelial ovarian cancer (EOC). In this cohort study with
Human exonuclease 1 (EXO1) activity characterization and its function on flap structures.
Keijzers, et al.
Bioscience Reports, 35 (2018)
Eugene Izumchenko et al.
DNA repair, 11(12), 951-964 (2012-10-16)
S(N)1 DNA methylating agents are genotoxic agents that methylate numerous nucleophilic centers within DNA including the O(6) position of guanine (O(6)meG). Methylation of this extracyclic oxygen forces mispairing with thymine during DNA replication. The mismatch repair (MMR) system recognizes these
Mahmoud El-Shemerly et al.
Nucleic acids research, 36(2), 511-519 (2007-12-01)
Nucleases play important roles in DNA synthesis, recombination and repair. We have previously shown that human exonuclease 1 (hEXO1) is phosphorylated in response to agents stalling DNA replication and that hEXO1 consequently undergoes ubiquitination and degradation in a proteasome-dependent manner.
Mu-Yan Cai et al.
Cell reports, 30(7), 2402-2415 (2020-02-23)
Cells deficient in ataxia telangiectasia mutated (ATM) are hypersensitive to ionizing radiation and other anti-cancer agents that induce double-strand DNA breaks. ATM inhibitors may therefore sensitize cancer cells to these agents. Some cancers may also have underlying genetic defects predisposing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service