Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503527

Sigma-Aldrich

Anti-PAR4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Coagulation factor II receptor-like 3, F2RL3, PAR-4, Proteinase-activated receptor 4, Thrombin receptor-like 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... F2RL3(9002)

General description

Anti-PAR4 Antibody detects endogenous levels of total PAR4 protein.
F2R-like thrombin or trypsin receptor 3/coagulation factor 2 receptor-like 3 (F2RL3) gene codes for protease-activated receptor 4 (PAR4) protein. PAR4 belongs to the G protein-coupled receptors (GPCRs) family. It is expressed at high levels in the lungs, pancreas, thyroid, testis, and small intestine. PAR4 gene is located on human chromosome 19p12. Anti-PAR4 antibody detects endogenous levels of total PAR4 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human PAR4.

Immunogen Range: 29-78

Application

Anti-PAR4 antibody produced in rabbit has been used in immunoblotting (1:1000).[1]

Biochem/physiol Actions

Protease-activated receptor 4 (PAR4) protein is involved in vascular inflammation. It is necessary for a stable thrombus as it mediates persistent thrombin signaling in platelets. PAR4 aids pro-inflammatory actions by interacting with the bradykinin B2 receptor. It is linked to renal function and the development of chronic kidney disease (CKD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Han et al.
Blood, 136(19), 2217-2228 (2020-06-24)
Protease-activated receptor 4 (PAR4) mediates sustained thrombin signaling in platelets and is required for a stable thrombus. PAR4 is activated by proteolysis of the N terminus to expose a tethered ligand. The structural basis for PAR4 activation and the location
Gamariel Rwibasira Rudinga et al.
International journal of molecular sciences, 19(2) (2018-02-15)
Cardiovascular diseases (CVDs) are currently among the leading causes of death worldwide. Platelet aggregation is a key cellular component of arterial thrombi and major cause of CVDs. Protease-activated receptors (PARs), including PAR1, PAR2, PAR3 and PAR4, fall within a subfamily
Dorothea M Heuberger et al.
Thrombosis journal, 17, 4-4 (2019-04-13)
Inflammatory diseases have become increasingly prevalent with industrialization. To address this, numerous anti-inflammatory agents and molecular targets have been considered in clinical trials. Among molecular targets, protease-activated receptors (PARs) are abundantly recognized for their roles in the development of chronic
Simeng Li et al.
International journal of molecular sciences, 20(22) (2019-11-14)
Protease-activated receptors (PARs) are a family of four GPCRs with a variety of cellular functions, yet the only advanced clinical endeavours to target these receptors for therapeutic gain to date relates to the impairment of platelet function for anti-thrombotic therapy.
Dian Ningtyas et al.
Frontiers in genetics, 11, 432-432 (2020-05-20)
The F2RL3 gene encoding protease activated receptor 4 (PAR4) contains a single nucleotide variant, rs773902, that is functional. The resulting PAR4 variants, Thr120, and Ala120, are known to differently affect platelet reactivity to thrombin. Significant population differences in the frequency

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service