Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503345

Sigma-Aldrich

Anti-ZNF397 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ZNF397, ZNF47, Zinc finger and SCAN domain-containing protein 15, Zinc finger protein 397, Zinc finger protein 47

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€347.00

€347.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€347.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 61 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF397(84307)

General description

Zinc finger protein 397 (ZNF397) belongs to the Cys2His2 group of the zinc-finger superfamily. It possesses a leucine-rich SCAN domain and nine Cys2His2 zinc fingers. The gene encoding this protein is localized on human chromosome 18q12.

Immunogen

The antiserum was produced against synthesized peptide derived from human ZNF397.

Immunogen Range: 10-59

Biochem/physiol Actions

Zinc finger protein 397 (ZNF397) has been shown to co-localize with centromere protein-A (CENP-A) during early prophase and interphase. It might be involved in transcriptional regulation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yimin Wu et al.
Gene, 310, 193-201 (2003-06-13)
We have identified two novel human SCAN domain genes ZNF396 and ZNF397, which are clustered within the region of chromosome 18q12. Three isoforms of ZNF396 transcript, 1.5, 2.5 and 3.9-kb, are expressed highly in liver. Four isoforms of ZNF397 transcript
S L Bailey et al.
Chromosoma, 117(4), 367-380 (2008-03-29)
The centromere is a complex structure required for equal segregation of newly synthesised sister chromatids at mitosis. One of the significant objectives in centromere research is to determine the complete repertoire of protein components that constitute the kinetochore. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service