Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503254

Sigma-Aldrich

Anti-VANGL1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

LPP2, Loop-tail protein 2 homolog, Strabismus 2, Van Gogh-like protein 1, Vang-like protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€347.00

€347.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€347.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 59 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VANGL1(81839)

General description

Anti-VANGL1 Antibody detects endogenous levels of total VANGL1 protein.
The gene VANGL1 (vang-like protein 1) is mapped to human chromosome 1p13. The encoded protein is a transmembrane protein and a component of the Wnt-PCP (planar cell polarity) pathway. The VANGL1 protein can interact with planar cell polarity (PCP) core proteins Disheveled, Prickle, and Frizzled, KAI1 (metastasis suppressor Kangai-1) protein and ITF (intestinal trefoil factor) protein.[1]

Immunogen

The antiserum was produced against synthesized peptide derived from human VANGL1.

Immunogen Range: 301-350

Biochem/physiol Actions

VANGL1 (vang-like protein 1) is associated with tumor progression in various cancers. In colorectal cancer, it enhances the angiogenesis. In mouse colon cells, overexpression of the VANGL1 gene causes tumorigenicity, invasiveness and adhesion to fibronectin.[2] VANGL1 is upregulated in colon, laryngeal, oral cavity squamous, gastric and hepatocellular cancer tissues.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon H Jiang et al.
Cell reports. Medicine, 2(12), 100475-100475 (2022-01-15)
We identify an intronic deletion in VANGL1 that predisposes to renal injury in high risk populations through a kidney-intrinsic process. Half of all SLE patients develop nephritis, yet the predisposing mechanisms to kidney damage remain poorly understood. There is limited
Gokhan Ozan Cetin et al.
Genetic testing and molecular biomarkers, 19(6), 283-287 (2015-04-16)
The Wnt planar cell polarity (PCP) pathway is one of the Wnt pathways which plays a critical role in cell proliferation and fate. The VANGL1 protein is one of Wnt-PCP pathway components. It is known that Wnt-PCP pathway has major
An unconventional KITENIN/ErbB4-mediated downstream signal of EGF upregulates c-Jun and the invasiveness of colorectal cancer cells.
Bae JA, et al.
Clinical Cancer Research, 20, 4115-4128 (2014)
Hyung-Hoon Oh et al.
Oncology reports, 35(1), 253-260 (2015-10-27)
Angiogenesis and lymphangiogenesis are involved in the dissemination of tumor cells from solid tumors to regional lymph nodes and various distant sites. KAI1 COOH-terminal interacting tetraspanin (KITENIN) contributes to tumor progression and poor clinical outcomes in various cancers including colorectal
Kyung-Hwa Lee et al.
Oncotarget, 6(5), 3240-3253 (2015-01-22)
KITENIN (KAI1 COOH-terminal interacting tetraspanin) promotes tumor invasion and metastasis in various cancers. This study assessed the association between KITENIN expression and advanced glioma grade in patients. In vitro assays revealed that KITENIN knockdown inhibited the invasion and migration of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service